NIST3131A
Magnesium (Mg) standard solution
NIST® SRM® 3131a
Select a Size
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 x 10 mL
manufacturer/tradename
NIST®
format
single component solution
General description
SRM 3131A_cert
SRM 3131A_SDS
Application
Features and Benefits
- Accompanied by a NIST-certified value of magnesium that is metrologically traceable to the SI unit of mass and expressed as a mass fraction.
- Expiration details, instructions for handling, storage and use are provided on the NIST certificate.
Other Notes
Magnesium (Mg)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service