Skip to Content
Merck
All Photos(1)

Key Documents

NIST980

Magnesium

NIST® SRM® 980, isotopic standard

Sign Into View Organizational & Contract Pricing

Select a Size

0.25 G
$1,180.00

$1,180.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.25 G
$1,180.00

About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,180.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

vapor pressure

1 mmHg ( 621 °C)

isotopic purity

 24Mg
 26Mg
 25Mg

autoignition temp.

950 °F

packaging

pkg of 0.25 g

manufacturer/tradename

NIST®

resistivity

4.46 μΩ-cm, 20°C

technique(s)

mass spectrometry (MS): suitable

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Magnesium-24 (24Mg), Magnesium-25 (25Mg), Magnesium-26 (26Mg)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Self-heat. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service