Skip to Content
Merck
All Photos(1)

Key Documents

NIST3114

Copper (Cu) standard solution

NIST® SRM® 3114

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$2,340.00

$2,340.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
50 ML
$2,340.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,340.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 50 mL

manufacturer/tradename

NIST®

format

single component solution

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of copper. A unit of SRM 3114 consists of 50 mL of an aqueous solution in a high-density polyethylene bottle sealed in an aluminized bag. The solution was prepared gravimetrically to contain a known mass fraction of copper. The solution contains nitric acid at a volume fraction of approximately 10 %, equivalent to an amount-of-substance concentration (molarity) of approximately 1.6 mol/L.For more information, plese refer to the SDS and COA.

SRM 3114_cert

SRM 3114_SDS

Other Notes

Certified for the analytes listed below.
Copper (Cu)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service