Skip to Content
Merck
All Photos(1)

Key Documents

NIST3126A

Iron (Fe) standard solution

NIST® SRM® 3126a

Synonym(s):

Iron Calibration Solution

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 10 ML
$2,240.00

$2,240.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 X 10 ML
$2,240.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,240.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of iron. A unit of SRM 3126a consists of five 10 mL sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of iron. The solution contains nitric acid at a volume fraction of approximately 10 %, equivalent to an amount-of-substance concentration (molarity) of approximately 1.6 mol/L. For more information, please refer to the COA and SDS.

SRM 3126A_cert

SRM 3126A_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Iron (Fe)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service