Skip to Content
Merck
All Photos(1)

Key Documents

NIST3107

Boron (B) standard solution

NIST® SRM® 3107

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$2,240.00

$2,240.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$2,240.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,240.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 50 mL

manufacturer/tradename

NIST®

format

single component solution

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of boron. A unit of SRM 3107 consists of 50 mL of an aqueous solution in a high-density polyethylene bottle sealed in an aluminized bag. For more information, please refer to the SDS and COA.

SRM 3107_cert

SRM 3107_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Boron (B)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service