Skip to Content
Merck
All Photos(1)

Key Documents

NIST3134

Molybdenum (Mo) standard solution

NIST® SRM® 3134

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 10 ML
$2,240.00

$2,240.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 X 10 ML
$2,240.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,240.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

Each unit of SRM 3134 contains five sealed borosilicate glass ampoules, each containing 10 mL of an acidified aqueous solution, gravimetrically prepared to have a known mass fraction of molybdenum.The solution includes approximately 10% volume fraction of hydrochloric acid, corresponding to an approximate molarity of 1.2 mol/L.

SRM 3134_cert

SRM 3134_SDS

Application

The SRM is intended as a primary calibration standard for the quantitative molybdenum determination.

Features and Benefits

  • The certified value of the SRM was determined by calculating the mass fraction values of mercury, which is obtained through two methods:
  1. Gravimetric preparation using high-purity molybdenum metal assayed by NIST
  2. Isotope-dilution cold-vaporinductively coupled plasma – mass spectrometry (ID-ICP-MS).
  • The certified value is metrologically traceable to the SI unit of mass fraction, expressed as micrograms per gram.
  • The NIST certificate is provided with period of validity, storage, handling, use, and maintenance instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Molybdenum (Mo)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service