Skip to Content
Merck
All Photos(1)

Key Documents

1.14761

Supelco

Iron Test, photometric

photometric, 0.0025-5.00 mg/L (Fe), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

250 TESTS
€126.00
1000 TESTS
€274.00

€126.00


Estimated to ship on29 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
250 TESTS
€126.00
1000 TESTS
€274.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€126.00


Estimated to ship on29 March 2025


Request a Bulk OrderRequest more information

Product Name

Iron Test, photometric, 0.0025-5.00 mg/L (Fe), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 1000 tests
sufficient for 250 tests (depending on package size)

specific analyte(s)

iron

measuring range

0.0025-5.00 mg/L (Fe)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Triazine)

storage temp.

15-25°C

General description

Method: photometric 0.0025 - 5.00 mg/l Fe Spectroquant®
This Spectroquant® Iron test allows the accurate quantification of iron ions in various waters. All iron ions are reduced to iron(II) ions. In a thioglycolate-buffered medium these react with a triazine derivative to form a red-violet complex that is determined photometrically.
MQuant® test strips can be used for pre-testing a sample in an economic and quick way to estimate the iron concentration and a possible dilution factor prior to using a quantitative method such as the Spectroquant® test.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spectroquant® Iron Test - a suitable alternative to ICP or AAS for determination of iron in drinking water, mineral water, groundwater, and spring water.

Protocols

Photometric determination subsequent to acid mineralisation

Related Content

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service