Skip to Content
Merck
All Photos(2)

Key Documents

1.11136

Supelco

Iron Test Kit, colorimetric

0.1-50 mg/L (Fe), MQuant®

Synonym(s):

Iron Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

200 TESTS
€224.00

€224.00


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
200 TESTS
€224.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€224.00


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Product Name

Iron Test, colorimetric, 0.1-50 mg/L (Fe), for use with MCOLORTEST®

specific analyte(s)

iron

Quality Level

measuring range

0.1-50 mg/L (Fe)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color-matching vessel 0.1 - 0.3 - 0.5 - 1.0 - 2.5 - 5.0 - 7.5 - 12.5 - 25 - 50 mg/l Fe MQuant®
The MQuant® Iron test (formally MColortest®) measures bivalent and trivalent iron in its dissolved form and also fresh colloidal e.g. in
  • Groundwater and surface water
  • Waters from aquaculture
  • Drinking water and mineral water
  • Industrial water
  • Wastewater and percolating water
  • Boiler and boiler feed water, cooling water
  • Food (e. g. wine) after appropriate sample pretreatment
This test is not suited for seawater.
We also recommend this as a pretest for the oxygen determination with MQuant® Oxygen 1.11107 to check for iron interferences in the sample.

This test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation. The color matching vessel is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.

Method
All iron ions are reduced to iron(II) ions by hydroxylammonium chloride. In a buffered medium these react with 2,2′-bipyridine to form a red complex. The iron concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color zones of a color matching vessel.
Without the addition of hydroxylammonium chloride (reagent Fe-1), the test measures only iron(II). so a differentialtion is possible.

Using the refill pack 1.08023 containing all the reagents, the original test kit can be used for a very long time, making this a particularly economical and ecological system. 
The test measures bivalent and trivalent iron in its dissolved form and also fresh colloidal iron(III)hydroxide in e.g. groundwater and surface water, drinking water, industrial water and wastewater, whereas a differentiation between iron(II) and iron(III) is possible.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service