Skip to Content
Merck
All Photos(1)

Key Documents

1.09713

Supelco

Nitrate Test, photometric

photometric, 0.10-25.0 mg/L (NO3-N), 0.4-110.7 mg/L (NO3-), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€213.00
250 TESTS
€380.00

€213.00


Estimated to ship on24 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
100 TESTS
€213.00
250 TESTS
€380.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€213.00


Estimated to ship on24 March 2025


Request a Bulk OrderRequest more information

Product Name

Nitrate Test, photometric, 0.10-25.0 mg/L (NO3-N), 0.4-110.7 mg/L (NO3-), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests
sufficient for 250 tests (depending on package size)

specific analyte(s)

nitrate

measuring range

0.10-25.0 mg/L (NO3-N)
0.4-110.7 mg/L (NO3-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (2,6-Dimethylphenol)

storage temp.

15-25°C

General description

Method: photometric, DMP 0.10 - 25.0 mg/l NO₃-N 0.4 - 110.7 mg/l NO₃⁻ Spectroquant®
The method is analogous to DIN 38405-9.
This Spectroquant® Nitrate Cell Test allows the accurate quantification of nitrate in water samples. Nitrate is an essential nutrient required for the growth of animals and plants. However, elevated levels of nitrate can be a health risk, in particular for pregnant women and infants. Therefore, nitrate levels in water sources are regulated in many countries.
Further fields of applications are the analysis of foods & beverages (e.g. whey) and of environmental and agricultural samples (e.g. soil).
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa Steliga et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-12)
Biodegradation is one of the most effective and profitable methods for the elimination of toxic polychlorinated biphenyls (PCBs) and total petroleum hydrocarbons (TPH) from the environment. In this study, aerobic degradation of the mentioned pollutants by bacterial strains Mycolicibacteriumfrederiksbergense IN53
Luis A Cueto et al.
Environmental science and pollution research international, 27(14), 15984-15993 (2019-05-06)
Phosphorus (P) is a nutrient for plant growth but also a pollutant in water bodies causing eutrophication. The source of P is mainly human and animal wastewater and runoffs from different land uses. The objective of the present study is

Articles

A step-by-step procedure for the photometric determination of Nitrate in effluents using Spectroquant® test kits and photometer for a precise and quantitative analysis.

Protocols

Learn about the photometric determination of nitrate ions in manure using Spectroquant® test kits and spectrophotometer.

Questions

  1. what is complete procedure to use kit

    1 answer
    1. Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service