Skip to Content
Merck
All Photos(1)

Key Documents

1.14406

Supelco

Manganese Test Kit, colorimetric

0.03-0.5 mg/L (Mn), MQuant®

Synonym(s):

Manganese Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

120 TESTS
€168.00

€168.00


Estimated to ship on24 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
120 TESTS
€168.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€168.00


Estimated to ship on24 May 2025


Request a Bulk OrderRequest more information

Product Name

Manganese Test, colorimetric, 0.03-0.5 mg/L (Mn), for use with MCOLORTEST®

specific analyte(s)

manganese

Quality Level

measuring range

0.03-0.5 mg/L (Mn)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with long test tubes 0.03-0.06-0.10-0.15-0.20-0.25-0.3-0.4-0.5 mg/l Mn MQuant®
The MQuant® Manganese test (formally MColortest®) measures bivalent manganese in its dissolved form.
It can be used for e.g.
  • Groundwater and surface water, seawater
  • Drinking water and mineral water
  • Spring water and well water
  • Boiler and boiler feed water, cooling water
  • Wastewater
  • Electroplating wastewater
  • Nutrient solutions for fertilization
  • Soils after appropriate sample pretreatment

Method
In alkaline solution manganese(II) ions react with an oxime to form a red-brown complex. The manganese concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Due to the long test tubes a sensitive measurement range can be covered.
Using the refill pack 1.18460 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system. 
The test measures bivalent manganese in its dissolved form. The manganese concentration can be measured semiquantitatively by visual comparison of the colour of the measurement solution with the colour fields of the color disk.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Respiratory system, spleen

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service