Skip to Content
Merck
All Photos(1)

Key Documents

1.00598

Supelco

Chlorine Test, photometric

photometric, 0.010-6.00 mg/L (Cl2), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

200 TESTS
€99.90
1200 TESTS
€277.00

€99.90


Estimated to ship on30 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
200 TESTS
€99.90
1200 TESTS
€277.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€99.90


Estimated to ship on30 March 2025


Request a Bulk OrderRequest more information

Product Name

Chlorine Test, photometric, 0.010-6.00 mg/L (Cl2), Spectroquant®

Quality Level

product line

Spectroquant®

usage

sufficient for 1200 tests (free chlorine)
sufficient for 200 tests (depending on package size)

specific analyte(s)

chlorine

measuring range

0.010-6.00 mg/L (Cl2)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Move DC
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (DPD)

storage temp.

15-25°C

General description

(free chlorine) Method: photometric, DPD 0.010 - 6.00 mg/l Cl₂ Spectroquant®
This Spectroquant® Chlorine test allows the accurate quantification of chlorine in various waters. Chlorinated disinfectants are still in common use all over the world, their major applications being in the chlorination of drinking water and swimming pools. In weakly acidic solution free chlorine reacts with dipropyl-p-phenylenediamine (DPD) to form a red-violet dye that is determined photometrically.
The method is analogous to EPA 330.5, APHA 4500-Cl2 G, and DIN EN ISO 7393-2.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about disinfection control and how to select the right disinfectant among the various options available in its methods and instrumentation based on photometry and reflectometry.

Protocols

Spectrophotometric determination of bromine with dipropyl-p-phenylenediamine (DPD) in drinking water, swimming-pool water, wastewater, and disinfection solutions in a range of 0.020 – 10.00 mg/L.

Preparation of a standard solution for free Chlorine according to DIN EN ISO 7393

Related Content

AQA Standard for free chlorine - Preparation of a standard solution for free chlorine

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service