Skip to Content
Merck
All Photos(1)

Key Documents

1.14688

Supelco

Crack Set 10C Pretreatment Kit

sufficient for 25 digestions of lead, cadmium, iron, copper, nickel, phosphorus (total) and zinc, Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
€172.00

€172.00


Estimated to ship on21 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
€172.00

About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

€172.00


Estimated to ship on21 May 2025


Request a Bulk OrderRequest more information

Product Name

Crack Set 10C, sufficient for 25 digestions of lead, cadmium, iron, copper, nickel, phosphorus (total) and zinc, Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 digestions of lead, cadmium, iron, copper, nickel, phosphorus (total) and zinc

specific analyte(s)

cadmium, copper, iron, lead, nickel, phosphorus, zinc

technique(s)

photometry: suitable

storage temp.

15-25°C

General description

for the digestion of lead, cadmium, iron, copper, nickel, phosphorus (total) and zinc 25 digestions Spectroquant®

Analysis Note

Application test: conforms

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service