Skip to Content
Merck
All Photos(1)

Key Documents

1.00607

Supelco

Ozone Test, photometric

photometric, 0.010-4.00 mg/L (O3), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

200 TESTS
€111.00
1200 TESTS
€338.00

€111.00


Estimated to ship on21 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
200 TESTS
€111.00
1200 TESTS
€338.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€111.00


Estimated to ship on21 May 2025


Request a Bulk OrderRequest more information

Product Name

Ozone Test, photometric, 0.010-4.00 mg/L (O3), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 1200 tests (depending on package size)
sufficient for 200 tests

specific analyte(s)

ozone

measuring range

0.010-4.00 mg/L (O3)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Move DC
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (DPD)

storage temp.

15-25°C

General description

Method: photometric, DPD 0.010 - 4.00 mg/l O₃ Spectroquant®This Spectroquant® Ozone test allows the accurate quantification in various waters. Ozone is a potent and useful oxidizing agent and strong biocide used in e.g. drinking water disinfection. In weakly acidic solution ozone reacts with dipropyl-p-phenylenediamine (DPD) to form a red-violet dye that is determined photometrically. The method is analogous to DIN 38408-5.

All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates. The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about disinfection control and how to select the right disinfectant among the various options available in its methods and instrumentation based on photometry and reflectometry.

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service