Skip to Content
Merck
All Photos(1)

Key Documents

1.18758

Supelco

Ozone Test Kit, colorimetric

0.15-10 mg/L (O₃), MQuant®

Synonym(s):

Ozone Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

300 TESTS
€281.00

€281.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
300 TESTS
€281.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€281.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Product Name

Ozone Test, colorimetric, 0.15-10 mg/L (O3), for use with MCOLORTEST®

product line

MColortest®

Quality Level

specific analyte(s)

ozone

measuring range

0.15-10 mg/L (O3)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric, DPD, with color-disk comparator 0.15 - 0.35 - 0.5 - 0.7 - 1.4 - 2.7 - 5.0 - 7.0 - 10 mg/l O2 MQuant®
The MQuant® Ozone test (formally MColortest®) measures the content of ozone in:
  • Drinking water
  • Wastewater
  • Process water
  • Disinfectant solutions
  • Swimming-pool water
This test is not suited for seawater.

Method
In weakly acidic solution ozone reacts with dipropyl-p-phenylenediamine (DPD) to form a red-violet dye. The ozone concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation. The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
Using the refill pack 1.18759 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

Application


  • Air Quality Monitoring: Ozone tests are crucial for monitoring air quality, particularly in urban and industrial areas where ozone pollution is a significant concern. These tests help in assessing the concentration of ozone in the atmosphere and its impact on public health and the environment. Recent research has reviewed various analytical methods applied for ozone gas detection, emphasizing their importance in environmental protection and regulatory compliance (Petruci et al., TrAC Trends in Analytical Chemistry, 2022).

  • Environmental Research: In environmental studies, ozone tests are used to investigate the sources and distribution of ozone in different ecosystems. These tests are vital for understanding the chemical reactions that produce ozone and its role in atmospheric chemistry. Advanced analytical techniques, such as portable deep-UV absorption sensors, are employed to measure ozone levels accurately, providing detailed insights into environmental ozone dynamics (Zheng et al., Trends in Environmental Analytical Chemistry, 2023).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service