Skip to Content
Merck
All Photos(2)

Key Documents

1.73635

Supelco

Move DC Hand-held Colorimeter

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€669.00

€669.00


Estimated to ship on26 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
1 UNIT
€669.00

About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

€669.00


Estimated to ship on26 March 2025


Request a Bulk OrderRequest more information

Product Name

Move DC,

product line

Spectroquant®

Quality Level

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move DC

storage temp.

15-25°C

General description

Spectroquant®
The Spectroquant Move Cl2/O3/ClO2/CyA/pH mobile colorimeter is suitable for the detection of five essential parameters of water analysis: chlorine, ozone, chlorine dioxide, cyanuric acid and pH. This handy, battery-operated instrument comes with a carrying case and is designed for in-process and field work.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about disinfection control and how to select the right disinfectant among the various options available in its methods and instrumentation based on photometry and reflectometry.

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service