Skip to Content
Merck
All Photos(1)

Key Documents

1.01744

Supelco

pH Test, photometric

photometric, pH range 6.4-8.8, Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

pH Cell Test, photometric, pH range 6.4-8.8, Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 280 tests

analyte functional class(es)

pH

technique(s)

photometry: suitable

pH range

6.4-8.8

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Move DC
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Indicator)

storage temp.

15-25°C

General description

Method: photometric pH 6.4 - 8.8 Spectroquant®

This Spectroquant® pH Cell Test allows the accurate determination of the pH value of aqueous samples in the range between pH 6.4 and 8.8.


Method applied: A non-chlorine-sensitive phenol red indicator solution changes color from yellow to red-violet depending on the pH. The resultant color is evaluated photometrically.


The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.


All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service