This product is a female Luer coupler (female-to-female) specified as compatible for male Luer to male Luer connections, so acts as a connector between two syringes with male Luer/Luer-lock tips. It is primarily listed as an accessory for LpDNPH/DNPH sampling cartridges/tubes used in air monitoring applications. An image has been provided below:
Fortfahren mit
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
packaging
pkg of 20 ea
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with male luer to male luer
General description
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description - | description pkg of 20 ea | description for use with detector tube | description - |
| technique(s) active air sampling: suitable | technique(s) active air sampling: suitable | technique(s) active air sampling: suitable | technique(s) active air sampling: suitable |
| compatibility for use with male luer to male luer | compatibility - | compatibility for use with detector tube | compatibility - |
| packaging pkg of 20 ea | packaging pkg of 20 ea | packaging - | packaging pkg of 1 ea |
| application(s) air monitoring | application(s) air monitoring | application(s) air monitoring | application(s) air monitoring |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
-
Is it a kind of connector between two syringes with Leur Lock.
1 answer -
Are these female Luer couplers sterile packed?
1 answer-
The female Luer couplers in Product No. 21015 are not sterile-packaged.