Merck
Z741758
4-liter waste bottle
Größe auswählen
1 EA
€ 326,00
€ 326,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 326,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
€ 326,00
Check Cart for Availability
Materialien
polycarbonate
Leistungsmerkmale
Comes with cap, tubing and fixtures.
Hersteller/Markenname
IBI ACCFL0009
Allgemeine Beschreibung
The 4L waste media bottle is set up to capture waste effluent from the IBI Flow Cell system. The bottle is made of polycarbonate and can be autoclaved. It comes complete with cap, containing fixtures and tubing to connect to the 3-channel or Convertible flow cell systems. Autoclavable & re-usable.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)