Merck
TMO312087-0008
Nalgene® narrow-mouth bottle
natural polypropylene copolymer, size 250 mL, case of 250 ea
Synonym(e):
Nalgene bottle, plastic bottle
Größe auswählen
250 EACH
€ 492,00
€ 492,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
250 EACH
€ 492,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
€ 492,00
Check Cart for Availability
Materialien
natural polypropylene copolymer
Verpackung
case of 250 ea
Hersteller/Markenname
Nalgene 312087-0008
Größe
250 mL
Allgemeine Beschreibung
Store and ship light-sensitive reagents with Thermo Scientific Nalgene Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)