Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
60 ea | Warenkorb auf Verfügbarkeit prüfen | € 410,00 |
Über diesen Artikel
€ 410,00
material
high-density polyethylene cap, square bottle, transparent PETG
joint
Joints threaded neck (38-430mm)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations: no, with cap
packaging
pack of 60 ea (2 xpack of 30)
manufacturer/tradename
Nalgene® 342020-0250
capacity
250 mL
General description
Features and Benefits
- Extensive validation documentation is available
- Produced from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG) that enhance reduced permeability to CO2/O2.
- Tamper evident shrink sleeves used along with moulded in graduations and neck ring.
- Non-pyrogenic and non-cytotoxic
- Animal derived component free (ADCF) material for bottle and closure material.
- Leakproof and transparent
- Devoid of costly washing, depyrogenation steps
- Prolonged storage of pH-sensitive liquids such as culture media
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description capacity 250 mL | description capacity 1000 mL | description capacity 2000 mL | description capacity 125 mL |
| sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated |
| capacity 250 mL | capacity 1000 mL | capacity 2000 mL | capacity 125 mL |
| feature with cap, graduations: no | feature with cap, graduations: no | feature graduations: no, with cap | feature graduations: no, with cap |
| material high-density polyethylene cap, transparent PETG , square bottle | material square bottle, transparent PETG , high-density polyethylene cap | material square bottle, high-density polyethylene cap, transparent PETG | material square bottle, transparent PETG , high-density polyethylene cap |
| packaging pack of 60 ea (2 xpack of 30) | packaging pack of 24 ea (2 xpack of 12) | packaging pack of 12 ea (2 xpack of 6) | packaging pack of 96 ea (4 xpack of 24) |
| joint Joints threaded neck (38-430mm) | joint Joints threaded neck (38-430mm) | joint Joints threaded neck (53B) | joint Joints threaded neck (38-430mm) |
Still not finding the right product?
Explore all of our products under Nalgene® media bottles, style 2020
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?