S1815
SecureSlip™ glass coverslip
L × W 12 mm × 12 mm
Synonym(e):
cover slips, coverslips, glass cover slips
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.22
Empfohlene Produkte
Sterilität
(sterile)
sterile
Verpackung
pkg of 15 coverslips per insert
pkg of 5 inserts
Hersteller/Markenname
Grace Bio-Labs
L × B
12 mm × 12 mm
Allgemeine Beschreibung
Coverslips are affixed to a thin microscopically transparent silicone base which secures the coverslip to culture vessels to prevent movement and cell damage during cultivation and assay procedures. During culture, cells may be imaged without interference from the silicone. Silicone perimeter surrounding each coverslip acts as a hydrophobic barrier to isolate liquids. Silicone backing removes easily (by peeling) for conventional mounting. Available as inserts which facilitate cultivation and batch processing in sets of up to 15 coverslips per plate or as individual silicone backed 12 mm × 12 mm coverslips.
Rechtliche Hinweise
SecureSlip is a trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)