Merck
HS120638
Reagent Reservoir
capacity 10 mL, polystyrene, non-sterile, pack of 100 (bulk packed)
Synonym(e):
Reagent container, Reagent storage
Größe auswählen
100 EACH
€ 65,10
€ 65,10
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EACH
€ 65,10
About This Item
UNSPSC-Code:
41121522
NACRES:
NB.01
€ 65,10
Check Cart for Availability
Materialien
V-bottom bottom
polystyrene
Sterilität
non-sterile
Verpackung
pack of 100 (bulk packed)
Hersteller/Markenname
Heathrow Scientific 120638
Kapazität
10 mL
Allgemeine Beschreibung
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Leistungsmerkmale und Vorteile
  • Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Disposable
  • Design Patent Pending
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. Does this reservoir fit a 12 multichannel pipette?
    1 answer
    1. The length of this reservoir is 2.8 inches and is not suitable for a 12 channel pipette. Please see the link below to review multichannel pipette reservoirs that may be of interest:
      https://www.sigmaaldrich.com/US/en/substance/texanreagentreservoirformultichannelpipettes1234598765