Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
100 ea | Warenkorb auf Verfügbarkeit prüfen | € 69,60 |
Über diesen Artikel
€ 69,60
material
V-bottom bottom, polystyrene reservoir
sterility
non-sterile
packaging
pack of 100 (bulk packed)
manufacturer/tradename
Heathrow Scientific 120642
reservoir capacity
25 mL
General description
Features and Benefits
- Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Disposable
- Design Patent Pending
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description reservoir capacity 25 mL, polystyrene reservoir, non-sterile, pack of 100 (bulk packed) | description capacity 100 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea) | description capacity 50 mL, PVC, sterile; electron beam irradiated, pack of 100 (packed as 5 ea) | description capacity 10 mL, polystyrene, non-sterile, pack of 100 (bulk packed) |
| material V-bottom bottom, polystyrene reservoir | material V-bottom bottom, polystyrene | material PVC , V-bottom bottom | material V-bottom bottom, polystyrene |
| packaging pack of 100 (bulk packed) | packaging pack of 200 (packed as 5 ea) | packaging pack of 100 (packed as 5 ea) | packaging pack of 100 (bulk packed) |
| reservoir capacity 25 mL | reservoir capacity - | reservoir capacity - | reservoir capacity - |
| sterility non-sterile | sterility sterile; electron beam irradiated | sterility sterile; electron beam irradiated | sterility non-sterile |
| manufacturer/tradename Heathrow Scientific 120642 | manufacturer/tradename Heathrow Scientific 120647 | manufacturer/tradename Heathrow Scientific 120646 | manufacturer/tradename Heathrow Scientific 120638 |
Still not finding the right product?
Explore all of our products under Reagent Reservoir
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?