Fortfahren mit
HS120643
Reagent Reservoir
capacity 50 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea)
Synonym(e):
Heathrow Scientific 120643, buffer reservoir, fluid chamber, fluid container, fluid vessel, liquid containment tank, liquid reservoir, liquid storage tank, sample reservoir, solution reservoir
zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
200 ea
Warenkorb auf Verfügbarkeit prüfen
€ 214,00
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121522
Capacity:
50 mL
Material:
V-bottom bottom, polystyrene
€ 214,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
V-bottom bottom, polystyrene
sterility
sterile; electron beam irradiated
packaging
pack of 200 (packed as 5 ea)
manufacturer/tradename
Heathrow Scientific 120643
capacity
50 mL
General description
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Features and Benefits
  • Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Disposable
  • Design Patent Pending
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
HS120647HS120646HS120638
capacity
50 mL
capacity
100 mL
capacity
50 mL
capacity
10 mL
material
V-bottom bottom, polystyrene
material
V-bottom bottom, polystyrene
material
PVC , V-bottom bottom
material
V-bottom bottom, polystyrene
packaging
pack of 200 (packed as 5 ea)
packaging
pack of 200 (packed as 5 ea)
packaging
pack of 100 (packed as 5 ea)
packaging
pack of 100 (bulk packed)
sterility
sterile; electron beam irradiated
sterility
sterile; electron beam irradiated
sterility
sterile; electron beam irradiated
sterility
non-sterile
manufacturer/tradename
Heathrow Scientific 120643
manufacturer/tradename
Heathrow Scientific 120647
manufacturer/tradename
Heathrow Scientific 120646
manufacturer/tradename
Heathrow Scientific 120638
Still not finding the right product?
Explore all of our products under Reagent Reservoir
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)