Merck
GBL505196
Grace Bio-Labs ONCYTE® NOVA nitrocellulose film slides
96 square pads, 6 mm diam.glass substrate
Synonym(e):
nitrocellulose slides, protein microarrays
Größe auswählen
1 EA
€ 232,00
€ 232,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 232,00
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.15
€ 232,00
Check Cart for Availability
Materialien
nitrocellulose (on glass)
Verpackung
pack of 1 ea
Hersteller/Markenname
Grace Bio-Labs 505196
Außenmaß
74 mm × 110 mm
Bindungsart
medium binding surface
Allgemeine Beschreibung
Grace Bio-Labs ONCYTE® NOVA nitrocellulose coated substrates are designed for SBS compliant multiwell plates and glass substrates for high throughput automated processing of microarrays.
Rechtliche Hinweise
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)