Merck
EPM1289-0050
Test Tube Rack, for Eppendorf® New Brunswick Innova® and Eppendorf® Excella® Shakers
size 22‑26 mm medium, holds 16 tubes
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.73
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Größe
22‑26 mm medium
360x80x129 mm
Leistungsmerkmale
holds 16 tubes
Hersteller/Markenname
Eppendorf® M1289-0050 (Global)
Allgemeine Beschreibung
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)