Fortfahren mit
Z336793
Workstation storage/dispenser box, PMMA
small
Synonym(e):
lab dispenser, pasteur pipette dispenser, tip dispenser, tube dispenser
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
NACRES:
NB.51
UNSPSC Code:
41121800
Manufacturer/tradename:
BRAND
manufacturer/tradename
BRAND
L × W × H
165 mm × 152 mm × 178 mm
General description
Ideal for storage and dispensing many small items used in the lab like disposable pipette tips, microcentrifuge tubes, pasteur pipette etc. The top cover is removable and the front cover is hinged.
Other Notes
BR131902 (India)
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)