Merck
CLS4395
Corning® Costar® HTS Transwell® cell culture plates
tissue-culture treated, sterile, reservoir, pack of 48 ea
Größe auswählen
48 EA
€ 418,00
€ 418,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
48 EA
€ 418,00
About This Item
UNSPSC-Code:
41122107
NACRES:
NB.13
€ 418,00
Check Cart for Availability
Sterilität
sterile
Verpackung
pack of 48 ea
Hersteller/Markenname
Corning 4395
Bindungsart
Tissue Culture (TC)-treated surface
Allgemeine Beschreibung
This allows cell culture on the membranes of all 96 well HTS Transwell plate types within a relatively large amount of medium. The tray provides optimum culture conditions across the whole plate and good buffering against pH changes and nutrient depletion. Trays contain a removable stabilizer which reduces medium sloshing during manipulation. Reservoir trays are used during the active cell growth and feeding phase prior to transferring the HTS Transwell system to a receiver plate which segregates the individual inserts for the final assay step. This reservoir tray is tissue culture treated and sterile for applications requiring co-cultures with the same or different cell types.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)