Merck
CLS3087
Corning® multiwell plates, plate lids and sealing mats
Thermowell GOLD 96 well sealing mat, rubber silicone septa mat, pierceable, non-sterile, 50/cs
Synonym(e):
96 well sealing mat, rubber silicone sealing mat, rubber silicone septa sealing mat
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien:
silicone
Materialien
silicone
Sterilität
non-sterile
Verpackung
pack of 10
case of 50
Hersteller/Markenname
Corning 3087
Allgemeine Beschreibung
  • Offers a unique piercible surface for easy access to reaction contents
  • Compatible with all Thermowell GOLD 96 well PCR microplates
  • Fully autoclavable and reusable for a more cost effective alternative to other sealing methods
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)