Wichtige Dokumente
1.17348
Millipore
MAS-100 Atmos® Gas Exhaust Adapter
for use with MAS-100 Atmos®
Größe auswählen
1173480001
€ 935,00
€ 935,00
Voraussichtliches Versanddatum20. Mai 2025
Weitere Informationen anfragen
Größe auswählen
Ansicht ändern
1173480001
€ 935,00
About This Item
UNSPSC-Code:
41103403
NACRES:
NA.74
€ 935,00
Voraussichtliches Versanddatum20. Mai 2025
Weitere Informationen anfragen
Materialien
stainless steel adapter (
)
Leistungsmerkmale
autoclavable
Hersteller/Markenname
MAS-100 Atmos®
Anwendung(en)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
Kompatibilität
for use with MAS-100 Atmos®
Allgemeine Beschreibung
The MAS-100 Atmos® Gas Exhaust Tube Adapter is an accessory of MAS-100 Atmos® Microbial Compressed Gas Sampler for safe removal of sampled gases to prevent intoxication and contamination of the environment.
The adapter has a customized gas exhaust tubing with a minimum inner diameter of 32 mm and a maximum length of 5 m. These are the conditions validated by the manufacturer.
The connector piece of the Gas Exhaust Tube is connected to the exhaust of the MAS-100 Atmos® instrument. The alternative tubing is fixed to the connector with the tri-clamp.
Anwendung
MAS-100 Atmos® Microbial Compressed Gas Sampler is used to sample pre-defined compressed gases like argon, carbon dioxide, compressed air, and nitrogen from pharmaceutical, cosmetic, food & beverage industry.
Leistungsmerkmale und Vorteile
Removal of potentially toxic gas out of the sampling environment
Rechtliche Hinweise
MAS-100 Atmos is a registered trademark of MBV AG