Merck
Z676020
Manual pipette racks
Manual pipette racks
1 of 1 reviewers received a sample product or took part in a promotion
clear acrylic rack
Synonym(e):
pipette holder, pipette stand, serological pipette rack
Größe auswählen
1 EA
€ 153,00
€ 153,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 153,00
About This Item
UNSPSC-Code:
41122801
NACRES:
NB.01
€ 153,00
Check Cart for Availability
Materialien
clear acrylic rack
Leistungsmerkmale
Holds 0.1 to 50 mL pipettes or cans
Hersteller/Markenname
Heathrow Scientific HS20611
B × T × H
291 mm × 88 mm × 401 mm
Allgemeine Beschreibung
Angled four shelved compartments keep your 0.1 to 50 mL pipettes or cans within easy reach, while preventing damage and clutter.
Crystal-clear, heavy-duty acrylic model allows viewing from any angle.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Questions
Reviews
1 of 1 reviewers received a sample product or took part in a promotion
Active Filters
  1. San Diego, CA
    • Reviews 3
    • Vote 1
    5 out of 5 stars.
    Very durable pipette rack!
    Sturdy acrylic serological pipette rack and angled just right to allow easy access to pipettes and tubes.