Merck
Z675482
Sterile economy sample spoon
20 mL
Synonym(e):
Economy Sample Spoon, Sterile Sample Spoon, dispensing tool
Größe auswählen
100 EA
€ 125,00
€ 125,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EA
€ 125,00
About This Item
UNSPSC-Code:
41122403
NACRES:
NB.35
€ 125,00
Check Cart for Availability
Beschreibung
PS
Verpackung
pack of 100 ea
Hersteller/Markenname
Bel-Art F36740-00005
Allgemeine Beschreibung
Moulded of lightweight and rigid, white high impact polystyrene. Individually packaged sterile spoons for one time use packed 100 per box.
  • Use to collect specific volumes of dry or liquid materials
  • Remains upright when filled or empty for use as a weighing boat
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)