Merck
Z675458
Sterile economy sample spoon
2.5 mL
Synonym(e):
dispensing tool
Größe auswählen
100 EA
€ 157,00
€ 157,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EA
€ 157,00
About This Item
UNSPSC-Code:
41122403
NACRES:
NB.35
€ 157,00
Check Cart for Availability
Beschreibung
PS
Hersteller/Markenname
Bel-Art F36740-00002
Allgemeine Beschreibung
Moulded of lightweight and rigid, white high impact polystyrene. Individually packaged sterile spoons for one time use packed 100 per box.
  • Use to collect specific volumes of dry or liquid materials
  • Remains upright when filled or empty for use as a weighing boat
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)