Merck
Z418420
Sigma-Aldrich
Aldrich® System 45 gas bubbler
About This Item
UNSPSC-Code:
41121800
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Höhe
135 mm
Allgemeine Beschreibung
Bubbler is designed to monitor gas flow to reactions. It can be used with argon or nitrogen to provide an inert atmosphere, or gaseous reagents can be added to reactions. Supplied with (2) Viton® O-rings and (2) quick-disconnect fittings.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
System 45 is a trademark of John Babashak
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)