Merck
Z106941
Sigma-Aldrich
Wheaton micro Soxhlet extractor
Synonym(e):
415500
Größe auswählen
1 EA
€ 655,00
€ 655,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 655,00
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.43
€ 655,00
Check Cart for Availability
Hersteller/Markenname
WHEATON®
Allgemeine Beschreibung
Consists of extractor, Allihn condenser, 25 mL Erlenmeyer flask, two Wheaton connections, two hose connections, and glass extraction thimble.
Leistungsmerkmale und Vorteile
  • Used for continuous extraction of analytes from a solid into an organic solvent
  • WHEATON® Connection eliminates the need for hooks, springs and claps common to standard ground joint designs
  • As the flask containing the solvent is heated, vapors rise in the larger outside tube, enter the water-cooled condenser, and liquate
  • When the liquid level in the extractor reaches the top of the bent tube, siphoning action returns the extract-enriched solvent to the flask
  • Wetted parts are made from either WHEATON® 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements or PTFE
Rechtliche Hinweise
WHEATON is a registered trademark of Wheaton Industries, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)