Merck
SLW3152/06A
Pyrex® Volumetric flask, heavy duty, class A, amber, USP/ISO/DIN tolerances,
capacity 10 mL
Synonym(e):
Pyrex, SciLabware 3152/06A
Größe auswählen
5 EA
€ 141,00
€ 141,00
Voraussichtliches Versanddatum11. April 2025
Größe auswählen
Ansicht ändern
5 EA
€ 141,00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
€ 141,00
Voraussichtliches Versanddatum11. April 2025
Materialien
borosilicate glass flask
Verbindung
joint ST/NS neck10/19
Leistungsmerkmale
accuracy class A (ISO 1042 and DIN 12664, United States Pharmacopoeia (USP),conformity ′H′ certified)
Verpackung
pkg of 5 ea
Hersteller/Markenname
SciLabware 3152/06A
Gesamt H
90 mm
Kapazität
10 mL
Allgemeine Beschreibung
  • Complies with ISO 1042 and DIN 12664
  • Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
  • Manufactured from borosilicate glass with an amber coating on exterior surfaces to facilitate use with light sensitive solutions
  • Each flask is laser etched with an individual serial number, batch code and date of manufacturer
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)