Merck
PL800GABUT-10
Plas-Labs replacement butyl gloves
for 2 piece glove set, size 10 (for 8 in. ports)
Größe auswählen
1 PAIR
€ 190,00
€ 190,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 PAIR
€ 190,00
About This Item
UNSPSC-Code:
46181504
NACRES:
NB.35
€ 190,00
Check Cart for Availability
Materialien
butyl rubber
Größe
10 (for 8 in. ports)
Leistungsmerkmale
reusable
for use with glove box sleeves
Verpackung
pack of 1 pair
Hersteller/Markenname
Plas-Labs 800GABUT-10
Allgemeine Beschreibung
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)