Merck
PL800GB-8
Plas-Labs butyl glove box gloves
for 1 piece glove set, size 8 (for 8 in. ports)
Größe auswählen
1 EACH
€ 716,00
€ 716,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EACH
€ 716,00
About This Item
UNSPSC-Code:
42132201
NACRES:
NB.35
€ 716,00
Check Cart for Availability
Größe
8 (for 8 in. ports)
Allgemeine Beschreibung
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)