Merck
BR386048
BRAND® Oxygen flask
volume 250-300 mL, with glass stopper NS 19/26
Synonym(e):
Bottles, Oxygen flasks, Winkler pattern
Größe auswählen
2 EA
€ 63,20
€ 63,20
Check Cart for Availability
Größe auswählen
Ansicht ändern
2 EA
€ 63,20
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
€ 63,20
Check Cart for Availability
Materialien
soda-lime glass
Verpackung
pack of 2 ea
Hersteller/Markenname
BRAND 386048
Kapazität
250-300 mL
Volumen
250-300 mL
Allgemeine Beschreibung
Soda-lime glass. For the determination of oxygen dissolved in water. The measured capacity is specified to ±0.01 mL. With labeling area. Solid, obliquely cut standard-ground glass stopper can be secured with a fastening clip. Each flask is calibrated with its own stopper, hence, stoppers and flasks are not interchangeable. Each flask and its stopper is marked with a unique matching identification number.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)