Merck
BR32820
BRAND® USP BLAUBRAND® class A measuring cylinder, tall form
volume 25 mL
Synonym(e):
BLAUBRAND®, Graduated Cylinders, USP
Größe auswählen
2 EA
€ 28,70
€ 28,70
Check Cart for Availability
Größe auswählen
Ansicht ändern
2 EA
€ 28,70
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
€ 28,70
Check Cart for Availability
Materialien
borosilicate glass
Leistungsmerkmale
DE-M marking
Hersteller/Markenname
BRAND 32820
Volumen
25 mL
Allgemeine Beschreibung
The BRAND® USP BLAUBRAND® class A measuring cylinder in tall form is crafted from high-quality Boro 3.3 glass, ensuring durability and precision. It complies with DIN EN ISO 4788 standards and is calibrated to contain (TC, In), making it suitable for accurate liquid measurement. The design features a spout for easy pouring and a hexagonal glass base for stability. Each measuring cylinder comes with a USP batch certificate. This measuring cylinder is an essential tool for laboratories requiring high accuracy and reliability in liquid measurement.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)