Merck
496332
Sigma-Aldrich
D-(−)-Isoascorbinsäure Natriumsalz Monohydrat
97%
Synonym(e):
D-Isoascorbinsäure Natriumsalz Monohydrat, Natriumerythorbat Monohydrat
About This Item
Empirische Formel (Hill-System):
C6H7NaO6 · H2O
CAS-Nummer:
Molekulargewicht:
216.12
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12162002
PubChem Substanz-ID:
NACRES:
NA.23
Assay
97%
mp (Schmelzpunkt)
165 °C (dec.) (lit.)
SMILES String
O.[Na+].OC[C@@H](O)C1OC(=O)C([O-])=C1O
InChI
1S/C6H8O6.Na.H2O/c7-1-2(8)5-3(9)4(10)6(11)12-5;;/h2,5,7-10H,1H2;;1H2/q;+1;/p-1/t2-,5-;;/m1../s1
InChIKey
NKESWEQTVZVSSF-DMWQRSMXSA-M
Allgemeine Beschreibung
Sodium D-isoascorbate monohydrate (NDAM) is an enantiomer of L-ascorbic acid that has an orthorhombic crystal structure. It is mainly used as an antioxidant in the preservation of food and beverages.
Anwendung
NDAM has non-linear optical properties with enhanced piezoelectric resonance peaks which can be useful for sensor based applications.
Lagerklassenschlüssel
11 - Combustible Solids
WGK
WGK 1
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Persönliche Schutzausrüstung
Eyeshields, Gloves, type N95 (US)
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1
1 of 1
The nonlinear optical properties of the monoclinic d-isoascorbic acid crystal.
Rao KR, et al.
Applied Physics B: Lasers and Optics, 122(11), 270-270 (2016)
A study of the piezoelectric resonance in metal organic NLO single crystals: Sodium D-isoascorbate monohydrate and Lithium L-ascorbate dihydrate.
AIP Conference Proceedings, 1728(1), 020157-020157 (2016)
Crystal growth and nonlinear optical properties of sodium D-isoascorbate monohydrate.
Rao KR, et al.
CrystEngComm, 15(33), 6594-6601 (2013)
Measurement and Correlation for the Solid Solubility of Antioxidants Sodium L-Ascorbate and Sodium Erythorbate Monohydrate in Supercritical Carbon Dioxide.
Wang T and Chang P
Journal of Chemical and Engineering Data, 60(3), 790-794 (2015)