Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3881

Corning® 96 Well Half-Area Microplate

flat bottom clear, black polystyrene, NBS (non-binding surface), bag of 25, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

black polystyrene
clear bottom
flat bottom clear

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 25
pkg of 25 inner package

manufacturer/tradename

Corning 3881

size

96 wells

well D

10.7 mm

well volume

190 μL

well working volume

25-125 μL

color

black

binding type

NBS (non-binding surface)

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 Well Half-Area Microplate has a 60% thinner bottom than conventional polystyrene plates and aid in lower background fluorescence.
  • Opaque black polystyrene walls.
  • Minimal well-to-well crosstalk during fluorescent assays.
  • Ideal for reducing protein and nucleic acid binding at low concentrations.
  • Applicable for both top and bottom reading instruments.
  • Recommended working volume of 25 to 125 μL.
  • Conforms to standard microplate footprint and dimensions.
  • Visual clarity for optical density or bottom read fluorescent/luminescent measurements.

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service