Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3693

Corning® 96 Well Half-Area Microplate

flat bottom, white polystyrene, bag of 25 inner package, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat bottom
round white wells
white bottom
white polystyrene
maximum volume:
190 μL
size:
96 wells
well D:
10.7 mm
working volume:
125 μL

material

flat bottom
round white wells
white bottom
white polystyrene

description

non-treated

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 25 inner package

manufacturer/tradename

Corning 3693

maximum volume

190 μL

size

96 wells

well D

10.7 mm

working volume

125 μL

color

white

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service