Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3992

Corning® 96 Well Half-Area Microplate

flat bottom, white polystyrene, NBS (non-binding surface), bag of 5, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

flat bottom
round wells
white bottom
white polystyrene

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 5
pkg of 5 inner package

manufacturer/tradename

Corning 3992

size

96 wells

well D

10.7 mm

well volume

190 μL

well working volume

25-125 μL

color

white

binding type

NBS (non-binding surface)

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 Well Half-Area Microplate is ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise.
  • White microplate enhances luminescent signals and has low background luminescence and fluorescence.
  • Designed to reduce well-to-well crosstalk during luminescent assays.
  • Nonbinding Surface (NBS) treated microplates have a non-ionic hydrophilic well surface, which makes them ideal for minimizing protein binding in homogeneous assays.
  • Conforms to standard microplate footprint and dimensions.
  • Flat bottom is best suited for specimen detection.
  • Lids are supplied separately.

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service