Key Documents
BR701371
BRAND® sealing film for microplates
self-adhesive, suitable for (for storage/fluorescence measurement), vinyl
Synonym(s):
Accessories, PCR, Plates, Sealing films
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
material
black
vinyl
vinyl film
sterility
non-sterile
feature
self-adhesive
packaging
pack of 50 ea
manufacturer/tradename
BRAND 701371
suitability
suitable for (for storage/fluorescence measurement)
General description
White and black films provide support for fluorescence and luminescence measurements and for microscopy. Weak signals are amplified and interference of light from outside is minimized.
Features and Benefits
  • Black, light-absorbing film for optimal fluorescence measurements
  • White, light-reflecting film for amplified luminescent signal
  • Storable down to -40 °C
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)