Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3880

Corning® 96 Well Half-Area Microplate

flat bottom clear, black polystyrene, bag of 25, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

material

black polystyrene
black polystyrene plate
clear bottom
flat bottom clear

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 25
pkg of 25 inner package

manufacturer/tradename

Corning 3880

maximum volume

190 μL

size

96 wells

well D

10.7 mm

working volume

125 μL

color

clear

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 Well Half-Area Microplate untreated polystyrene surface (hydrophobic) enables biomolecules binding.
  • Opaque black polystyrene walls.
  • Without lids; recommended working volume of 25 to 125 μL
  • Lower background fluorescent and reading below 340nm.
  • Minimal well-to-well crosstalk during fluorescent assays.
  • Applicable for both top and bottom reading instruments.
  • Binding capacity of approximately 100 to 200ng IgG/cm².
  • Standard microplate footprint and dimensions format.
  • Clear polystyrene enables visual clarity in fluorescent and luminescent measurements.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service