Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3697

Corning® 96 Well Half-Area Microplate

flat bottom clear, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

clear wells
flat bottom clear
round wells

sterility

sterile

feature

lid
skirt

packaging

bag of 20

manufacturer/tradename

Corning 3697

size

96 wells

well D

10.7 mm

well volume

190 μL

well working volume

25-125 μL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 Well Half-Area Microplate are ELISA Microplates with flat bottoms
  • Sterilized by gamma radiation and nonpyrogenic
  • TC-treated for optimal cell attachment
  • Nonreversible lids with condensation rings to reduce contamination
  • Individual alphanumeric codes provide well identification

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service