Skip to Content
MilliporeSigma
All Photos(1)

Documents

4SB3

from Mouse x Mouse Hybridoma, 92030610, Reactivity Anti-interferon gamma, human )

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

P3X63Ag8

Reactivity

Anti-interferon gamma, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

Cell Line Origin

Mouse P3X63Ag8 x Mouse BALB/c spleen

Cell Line Description

This weakly neutralising antibody is directed against natural human IFN gamma and cross-reacts with recombinant IFN gamma (non-glycosylated, E.coli derived). There is no cross-reaction with IFN alpha or beta.

Immunogen

Human leukocyte-derived interferon gamma

Application

Immunoassays

Culture Medium

RPMI 1640 + 2 mM glutamine + 10% Fetal Bovine Serum (FBS) (heat inactivated)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service