Skip to Content
MilliporeSigma
All Photos(1)

Documents

MDBK Cell Line bovine

90050801, bovine kidney, Epithelial-like

Synonym(s):

MDBK (NBL-1) Cells, Madin Darby Bovine Kidney Cells, NBL-1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102

product name

MDBK Cell Line bovine, 90050801

biological source

bovine kidney

growth mode

Adherent

karyotype

Modal no. 51; hypodiploid

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Bovine kidney

Cell Line Description

Derived from the kidney of an apparently normal, adult Bos taurus. Also known as MDBK (NBL-1).

Application

MDBK cell line has been used to study the expression of Eimeria bovis-derived antigens on the host cell surface membrane in vitro. It has also been used to study the ubiquitination and degradation of signal transducer and activator of transcription 1 and 2 (STAT1 and STAT2) by the V proteins of the paramyxoviruses simian virus 5 and human parainfluenza virus type 2.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS) or 10% Horse Serum (HS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service