Skip to Content
MilliporeSigma
All Photos(1)

Documents

MT4/E4

92030608, Reactivity: Anti-Interferon α-2, human and some related subtypes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

P3X63Ag8

Reactivity

Anti-Interferon α-2, human and some related subtypes

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P3-X63-Ag8 x Mouse BALB/c spleen

Cell Line Description

MT4/E4 is a very strong neutralising monoclonal antibody with a wide range of neutralising monoclonal activities against homologous human IFN-α-2. The heterologous IFN-α-1 is weakly bound but also shows cross-reactive neutralisation. There is no cross-reaction with human IFN-ß or IFN-gamma.

Immunogen

Recombinant human interferon alpha-2

Application

Cytokine assays, studies of anti-viral activity of interferon, neutralisation of interferon-α-2 activity.

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS) (heat inactivated)

Subculture Routine

Maintain cultures between 5-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service